Learn More
Agilent Technologies 5x Capillary Conditioning Solution, 100 ml
Supplier:  Agilent Technologies DNF4750100
Encompass
5x Capillary Conditioning Solution, 100 mL. Solution necessary for conditioning and restoring capillary array performance. 5x concentration. For use with the Fragment Analyzer and ZAG DNA Analyzer systems.
Catalog No. NC1061046