Learn More
Dino Lite USB Digital Microscope AM7515MT8A - 5MP, 700x - 900x Optical Magnification, Measurement, AMR, Axial Lighting
Supplier: Dino Lite AM7515MT8A
The AM7515MT8A is a 5MP Dino-Lite Edge Series microscope that has a magnification range of 700x- 900x and built-in axial lighting. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. AMR displays the current magnification on screen with the included software for Windows or Mac OS. Through the use of Flexible LED Control (FLC) using the included software, this model is capable of switching between (or mixing) brightfield and darkfield illumination, revealing details that are very difficult to see under normal light.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.