Learn More
Bel-Art™ 5mm O.D. Thin Wall Precision NMR Sample Tube
Features 15% more sample volume than the standard wall Precision tube, and 25% more sample volume than the Economy NMR tube.
Supplier: Bel-Art™ 666050581
Description
NMR tubes are thin glass walled tubes used to contain samples in nuclear magnetic resonance spectroscopy. It is important that the tubes are uniformly thick and well-balanced to ensure that they spin at regular rates (i.e., they do not wobble), within the NMR spectrometer. NMR tubes are used primarily with small samples dissolved in an appropriate solvent.
Specifications
NMR Sample Tube | |
5 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.