Learn More
Revvity Health Sciences Inc Anti-human IgG (Fc specific) AlphaLISA Acceptor Beads 5mg
Supplier: Revvity Health Sciences Inc AL103M
These beads can be used in conjunction with Alpha Donor beads to create AlphaLISA no-wash immunoassays for: Analyte detection assays Biomarker detection assays Antibody detection assays Antibody binding studies Protein-protein interaction assays Other immunoassays In a typical AlphaLISA assay, 1 mg of Acceptor beads is sufficient to run 1,000-2,000 wells using a 50 ¿L reaction volume. Features: No-wash steps, no separation steps Ease-of-use: few addition steps, fast assay development Broad range of affinities: detect strong or weak interactions, from pM to mM affinity Distance: measure very large protein or antibody complexes ¿ spanning up to 200 nm or more High avidity: multiple binding sites on each bead enables use of nanomolar concentrations of antibodies or proteins, as well as use of low affinity binders AlphaScreen and AlphaLISA are bead-based assay technologies used to study biomolecular interactions in a microplate format. The acronym Alpha stands for amplified luminescent
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.