Learn More
Chemglass Life Sciences 50mL Flask, Storage, 14/20 Outer Joint, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF052201
50mL Flask, Storage, 14/20 Outer Joint, Airfree, Schlenk. Round bottom storage flask with a standard taper outer joint on the sidearm. The 50, 100 and 250mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 500 and 1000mL sizes have a 0-8mm valve size.