Learn More
Chemglass Life Sciences 50mL Funnel, Cold Addition, 24/40 Top Outer, 24/40 Lower Inner, 325mm OAL, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF055202
50mL Funnel, Cold Addition, 24/40 Top Outer, 24/40 Lower Inner, 325mm OAL, Airfree, Schlenk. Used in the preparation of block copolymers or any chemical synthesis requiring high vacuum techniques. Funnel features a large diameter ring seal between the coolant container & the addition funnel body. The open cup design of the coolant chamber allows for easy addition of coolant, particularly dry ice chunks. Constructed using grease-free 0-4mm Chem-Vac™ Chem-Cap valves on the pressure equalizing arm & lower inner joint. Positioning of the valves permits the funnel contents to be kept under inert conditions prior to use or during loading. A 14/20 outer joint is sealed on the funnel body above the coolant chamber for syringe sampling of the funnel contents. Funnel has a drain tube on the rear of the coolant chamber. Designed & developed by William J. Swatos & Barton J. Schober, Department Of Materials Science & Engineering, Polymer Science Program, Penn State University
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.