Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 6 25 kD MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
$220.00
Specifications
MWCO | 25 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1 meter of Spectra/Por 6 Dialysis Tubing
- 1 Standard Spectra/Por Closure (for buoyancy)
- 1 Weighted Spectra/Por Closure (for orientation)
- 5 Tubing Opening Picks
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
08-700-174
|
Spectrum™
132552T |
2.5 mL/cm vol./length | 18 mm |
Each for $220.00
|
|
||||
08-700-175
|
Spectrum™
132554T |
3.7 mL/cm vol./length | 22 mm |
Each for $220.00
|
|
||||
08-700-173
|
Spectrum™
132550T |
0.45 mL/cm vol./length | 7.5 mm |
Each for $220.00
|
|
||||
Description
- Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
- Membrane packaged wet in preservative solution of 0.05% sodium azide
- Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
- Include a pair of closures and convenient tubing opening picks
Specifications
25 kDa | |
1 m Tubing/Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title