Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Specifications
Quantity | 1500/Pk. |
---|---|
Material | Acetyl Resin |
Product Type | Tissue Processing Cassette in Hopper tube |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-222-076
|
Cellpath
EAD010504H |
Gray | N/A | N/A | |||||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
22-222-088
|
Cellpath
EAD012004H |
Turquoise | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-086
|
Cellpath
EAD011804H |
Dark Green | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-087
|
Cellpath
EAD011904H |
Lavender | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-085
|
Cellpath
EAD011604H |
Dark Blue | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-084
|
Cellpath
EAD011504H |
Brown | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-083
|
Cellpath
EAD011204H |
Tan | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-081
|
Cellpath
EAD011004H |
Red | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-080
|
Cellpath
EAD010904H |
Yellow | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-082
|
Cellpath
EAD011104H |
Peach | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-078
|
Cellpath
EAD010704H |
Orange | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-079
|
Cellpath
EAD010804H |
Pink | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-077
|
Cellpath
EAD010604H |
Blue | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-075
|
Cellpath
EAD010404H |
Green | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-074
|
Cellpath
EAD010204H |
White | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Pre-loaded into hoppers for use with automated and carousel type cassette printers
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
- Available in 15 stock colors which color code different types of workload or applications
Specifications
1500/Pk. | |
Acetyl Resin | |
Tissue Processing Cassette in Hopper tube |