Learn More
Cellpath System II Hex™ Tissue Processing Cassettes
To hold tissue specimens during embedding, processing, and storage. CellPath System II Hex Tissue Processing Cassettes feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
$172.20
Specifications
Quantity | 1000/Pk. |
---|---|
Material | Acetyl Resin |
Product Type | Tissue Processing Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-222-059
|
Cellpath
EAD010203A |
White |
Pack of 1000 for $172.20
|
|
|||||
22-222-068
|
Cellpath
EAD011203A |
Tan | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-067
|
Cellpath
EAD011103A |
Peach | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-066
|
Cellpath
EAD011003A |
Red | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-065
|
Cellpath
EAD010903A |
Yellow | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-073
|
Cellpath
EAD012003A |
Turquoise | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-064
|
Cellpath
EAD010803A |
Pink | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-072
|
Cellpath
EAD011903A |
Lavender | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-071
|
Cellpath
EAD011803A |
Dark Green | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-070
|
Cellpath
EAD011603A |
Dark Blue | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-069
|
Cellpath
EAD011503A |
Brown | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-063
|
Cellpath
EAD010703A |
Orange | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-062
|
Cellpath
EAD010603A |
Blue | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-060
|
Cellpath
EAD010403A |
Green | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
22-222-061
|
Cellpath
EAD010503A |
Gray | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Available in 15 stock colors which color code different types of workload or applications
Specifications
1000/Pk. | |
Acetyl Resin | |
Tissue Processing Cassette |