Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Labs 3.5 - 5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
$230.00 - $367.00
Specifications
MWCO | 3.5 to 5 kDa |
---|
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
08-700-120
|
Spectrum™
131204T |
3.1 mL/cm vol./length | 20 mm |
Each for $290.00
|
|
||||
08-700-118
|
Spectrum™
131192T |
0.32 mL/cm vol./length | 6.4 mm |
Each for $230.00
|
|
||||
08-700-119
|
Spectrum™
131201T |
1.8 mL/cm vol./length | 15 mm |
Each for $243.00
|
|
||||
08-801-249
|
Spectrum™
131198T |
0.79 mL/cm vol./length | 10 mm |
Each for $367.00
|
|
||||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
3.5 to 5 kDa |
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'