Learn More
Chemglass Life Sciences 500mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Coarse Frit, GL 32 Thread
SDP
Supplier:  Chemglass Life Sciences CG186615
Encompass_Preferred
500mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Coarse Frit, GL 32 Thread. Similar to CG-1864, but with the addition of a lower standard taper inner joint to allow collection into a vessel having a 24/40 outer joint. COARSE porosity fritted glass resin support. A serrated hose connection is sealed on the inner joint for connection to a vacuum source. The “T”-Bore stopcock has an 8mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8mm at the middle ring.
Catalog No. CG186615