Learn More
Invivogen Control for 5'ppp-dsRNA, 100 ug
Supplier: Invivogen TLRL3PRNAC100
5’ppp-dsRNA Control is a negative control for 5’ triphosphate double-stranded RNA (5’ppp-dsRNA), a synthetic ligand for the retinoic acid-inducible protein I (RIG-I, also known as Ddx58). RIG-I is a cytoplasmic RNA helicase that belongs to the RIG-I-like receptors (RLRs) family and triggers an antiviral immune response by the activation of type-I interferons (IFN-alpha and -beta).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.