Learn More
Invivogen Imiquimod VacciGrade, 5 mg
Supplier: Invivogen VACIMQ
The imidazoquinoline compound Imiquimod is a guanosine derivative and agonist for TLR7. This TLR7 agonist, originally developed as type I IFN inducers, is an effective adjuvant by activating dendritic cells (DCs) and B cells to induce cytokines optimal for Th1 cell immunity, and antibody production.