Learn More
Labelmaster Inc 5 GALPLASTIC DRUM WITH RIEKE S
Supplier:  Labelmaster Inc KH5020R1
Encompass_Preferred
5 gal HDPE High Density Polyethylene with 70mm Rieke screw cap. Lining HDPE
Catalog No. NC1648229