Learn More
BioSpec Products 48 HOLE ALUM VIAL HOLDER
Supplier: BioSpec Products 702ALU
Forty-eight hole, solid aluminum vial rack for 1.5 or 2.0ml microvials. This solid metal rack with standard microplate sized footprint can be pre-chilled in a refrigerator or freezer to aid in sample cooling. Can be used with Mini-96. 1 ea