Learn More
Edmund Optics 46MM SOFT JAW PLIERS
Supplier: Edmund Optics NT59900
The slip joint design utilized in the 46mm soft jaw pliers is ideal for handling optics, camera-lens assemblies, and other delicate components. Jaws feature screw-in plastic cushions to protect fine knurling and delicate finishes.