Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ 45 Gallon Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 45 Gallon Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet is specially designed for safely storing acids and corrosives in laboratories.
Supplier: Justrite™ 894522
Description
Justrite blue steel safety cabinets for corrosives have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet. Self-close door(s) shuts and latches automatically when a fusible link melts at 165°F (74°C) under fire conditions. Unique, concealed self-closing mechanism offers obstruction-free access to contents.Specifications
Self-close | |
2 | |
2 | |
457 x 1092 x 1651 mm <18 x 43 x 65 in.) | |
Blue |
45 gal. (170 L) | |
Cold-rolled Steel (18 ga) | |
370 x 1003 x 1537 mm (14.562 x 39.5 x 60.5 in.) | |
Steel (CR) | |
NFPA, OSHA, IFC, FM Approved |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction