Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4,5-Dimethoxy-o-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2'-dithiobis(1-aminonaphthalene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).
Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.
Notes
Incompatible with oxidizing agents.
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).
Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | ∽265°C (decomposition) |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 3714004 |
Solubility Information | Soluble in warm methanol,dimethyl sulfoxide and water. |
IUPAC Name | 4,5-dimethoxybenzene-1,2-diamine dihydrochloride |
Formula Weight | 241.12 |
Percent Purity | 98% |
Chemical Name or Material | 4,5-Dimethoxy-o-phenylenediamine dihydrochloride |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction