Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Fit Test Solution FT-12, Sweet
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.
Supplier: 3M™ 7100004636
Description
- Sweet taste fit test solution for fit testing
- Used with the 3M™ Qualitative Fit Test Apparatus FT-10 and 3M™ Training and Fit Testing Case FT-20
- Can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
- These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
Individual 1.9 oz. (55mL) bottles of sensitivity solution or fit test solution also available.
Compliance
Meets the performance criteria for fit testing respirators under OSHA 29 CFR 1910.134, Appendix A.
Specifications
Sweet Solution Refill | |
55 mL | |
Qualitative fit test kit | |
6/Cs. |
1.9 oz. | |
OSHA 29 CFR 1910.134, Appendix A | |
FT-12 | |
Sweet solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction