Learn More
MEDPLUS SERVICES USA 3M ATTEST RAPID READOUT BIOL
Supplier: MEDPLUS SERVICES USA MMM 1292
3M Attest Rapid Readout Biological Indicators for 250F/121C Gravity and 270F/132C Vacuum Assisted Steam Sterilizers