Learn More
3M™ 6000 Series Drop-Down Half Facepiece
Designed for workers who need to communicate frequently
Supplier:  3M™ 6300DD
 View more versions of this product
Catalog No. 19-041-420
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Requires : Facepiece requires compatible cartridges and/or filters. (See separate listings.)
This item is not returnable. View return policy
Description
Description
  • Head cradle adjusts to the individual worker's head size
  • Head harness system allows facepiece to drop to the chest without user having to remove hardhat
  • Durable straps hold their elasticity for long wear
  • Upper strap adjustment is on the head cradle so the strap ends do not interfere with worker's vision
  • Exhalation valve cover directs air downward to help prevent fogging of the faceshield
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
Can be used with Series 2000 or 7093 filters or Series 6000 cartridges. Contact your Fisher Safety Sales Representative for compatibility information related to various applications.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
NIOSH 42CFR84
Thermoplastic Elastomer
Large
Filter 2000 series, filter 7093, p100 or cartridge 6000 series
Bayonet
Drop Down Half Facepiece Respirator
Demolition, facility maintenance, sanding
6300DD
SDS
Documents
Documents
Product Certifications
Accessories