Learn More
MEDPLUS SERVICES USA 3M 116 ATTEST BIOLOGICAL INC
Supplier: MEDPLUS SERVICES USA MMM 116
• 3M Attest Incubators provide optimal conditions for accurate readout of 3M Attest Biological Indicators • 15-well capacity. • Dry block pre-set to 56˚C incubation temperature • Manage record-keeping with web app and your internet connected PC