Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Anatomy Posters
SCI_ED

Visual aids to enhance anatomy lessons.

Supplier:  3B Scientific™ VR1621L

 View more versions of this product

Catalog No. S90773


Add to Cart

Description

Description

  • Printed on premium glossy (200g) paper
  • With two-sided lamination (125 micron) and metal eyelets
  • Dimensions: 20W × 26 in.H (50.8 × 66.0cm)

Specifications

Specifications

Spinal Nerves Chart
26 in.
20 in.
Important spinal nerves and nerve sections are detailed
Science
66 cm
50.8 cm
Great addition to any doctor's office or classroom
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit