Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One 384-well, Advanced TC Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Supplier: Greiner Bio-One 781986
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Non-cytotoxic
- Improved cell adherence
- Permits usage of serum-reduced or serum-free media
- Optimal cultivation conditions for sensitive cells
Specifications
Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic | |
With Lid | |
16 x 24 | |
Clear | |
384 | |
Sterile | |
Rounded Square | |
3000 x g (swinging-bucket rotor) | |
8 | |
128 x 85 x 15 mm |
Standard | |
10 mm2 | |
Not Autoclavable | |
Polystyrene | |
15 to 110 μL | |
Advanced TC | |
Yes | |
32 | |
Yes |
Safety and Handling
ShelfLife : 2 Years after month of production
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction