Learn More
Carver Inc 3619 13 MM PELLET DIE FOR KBR
Supplier: Carver Inc 3619
13mm Evacuable Pellet Die for KBr Pellets, Complete with plunger, cylinder, base plug, ejector & (2) Anvils. 440 Stainless steel, Max load 18,000 lbs