Learn More
Applied Biosystems™ 310 Genetic Analyzer Matrix Standards Kit with Manual and Software, dRhodamine
A premade kit containing matrix standards for use with dRhodamine sequencing chemistries
Manufacturer:  Applied Biosystems™ 403047
Encompass
Catalog No. 40-304-7
Add to cart
Includes: Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Includes Manual and Software
Description
Description
The kit contains matrix standards for use with dRhodamine sequencing chemistries on the 310 Genetic Analyzer.
Order Info
1 Kit
Specifications
Specifications
Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Includes Manual and Software.
Sequencing
310 Genetic Analyzer Matrix Standards Kit
Kit
dRhodamine 110, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA
1 Kit