Learn More
Chemglass Life Sciences Reaction Vessel Lid, 7-Neck, 300mm
Supplier: Chemglass Life Sciences CG1968A01
Reaction Vessel Lid, 7-Neck, 300mm. 7-Neck reaction vessel lid for 300mm flanged vessels. Lid has a CN 45/50, 5-SN 45/50, and 1-SN 80mm Duran o-ring flange angled for powder addition.PLEASE NOTE: 80MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.*** PLEASE NOTE: THE ANGLED POWDER ADDITION PORT IS NOW 80MM AND NOT 100MM AS WAS SUPPLIED WITH PREVIOUS VERSION ***
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.