Learn More
Nuaire Inc TankGuard Automatic CO2 Incubator Tank Switch
Supplier: Nuaire Inc NU1550
The TankGuard™ automatic tank switch has been designed to provide reliable service to perform the critical backup gas (i.e. CO2, N2) tank switch for a tissue cell culture incubator. The TankGuard™ tank switch continuously monitors and indicates the selected source gas tank and the status of the non-selected or secondary source gas tank. If the selected source gas tank pressure falls below 8 PSI, the selected source internal pressure switch will detect the low pressure and automatically change sources. Includes: Power Cord-Tubing with in-line filter-2 x tubing lines-4 x tubing crimps-2 x plastic T valves
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.