Learn More
Cole-Parmer Koflo 3/8-24PPT Mounted in-line mixer; 3/8" tube OD, 24 elements, 10/pack
Supplier:  Cole-Parmer 0466814
Encompass
Mounted mixers can be used with any compression fitting with the same tubing OD.
Catalog No. NC1359146
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.