Learn More
MSC Hertel E2040024 3/8", 1-1/2" Length of Cut, 3/8" Shank Diam, 3-1/4" Overall Length, 4 Flute, Cobalt Square End Mill Single End, Uncoated, Spiral Flute, 30° Helix, Centercutting, Right Hand Cut, Right Hand Flute
Supplier:  MSC 41114265
Encompass
Hertel E2040024 3/8", 1-1/2" Length of Cut, 3/8" Shank Diam, 3-1/4" Overall Length, 4 Flute, Cobalt Square End Mill Single End, Uncoated, Spiral Flute, 30° Helix, Centercutting, Right Hand Cut, Right Hand Flute
Catalog No. NC1192847