Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorophenol, MilliporeSigma™ Supelco™

3,5-Dichlorophenol belongs to the class of substituted phenol compounds, which are widely used in industrial applications such as synthesis of pesticides, dyes, drugs, plastics, etc. Due to their extensive usage, they are commonly found in environmental samples as pollutants.

Manufacturer:  MilliporeSigma Supelco 442378

Catalog No. 11-102-0256


Add to cart

Description

Description

Agriculture and Environmental
Specifications

Specifications

591-35-5
MFCD00002259
Neat
Ampule
233°C
Organic Pollutant Standards
Cl2C6H3 OH
163.00
Analytical Standard
65°C to 68°C
1000 mg
Safety and Handling

Safety and Handling

RTECSNumber : SK8820000

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit