Learn More
Chemglass Life Sciences Beads, Borosilicate Glass, 2mm, Approx. 46,000 Pcs. Per Lb.
Supplier: Chemglass Life Sciences CG110101
Beads, Borosilicate Glass, 2mm, Approx. 46,000 Pcs. Per Lb. Solid borosilicate spheres can be used as distillation column packing, boiling stones or to provide support for chromatography columns used in gel filtrations. Beads have excellent chemical resistance. Packed in boxes containing one pound.