Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
29-Series Pop Beads
Create models of DNA strand, DNA replication, RNA conversion, cell organelles, ribosomes, mitochondria and a nucleus.
Supplier: Science First 6380324
Description
- Formed by proprietary molds to meet demanding specifications
- Versatile
- Bead diameter: 13mm (0.5 in.)
- Economical
- Easy to use
Specifications
Purple | |
0.5 in. | |
Create model of DNA strand, DNA replication, RNA conversion, cell organelles, ribosomes, mitochondria and nucleus |
Pop Bead, 3 Way | |
1.3 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction