Learn More
Miles Scientific 25ul Syringe for SpotON or TLC Autospotter (each)
Supplier: Miles Scientific A8732
Custom SGE brand TLC syringes with blunt PTFE tipped needles.Custom SGE brand TLC syringes with blunt PTFE tipped needles.These special needles minimize sample creep back and enhance reproducibility. The syringes also feature PTFE plunger tips to help eliminate the problem of metal-to-glass contamination.