Learn More
Ace Glass, Inc. Trap, vacuum, 25mm body diameter, 200mm body length, 10mm OD inner & side tube
Supplier: Ace Glass, Inc. 875205
Trap, vacuum, 25mm body diameter, 200mm body length, 10mm OD inner & side tube