Learn More
Chemglass Life Sciences 250mL Storage Flask, Cajon, Airfree, Schlenk, 0-4mm Valve, 160mm OAH
Supplier: Chemglass Life Sciences AF009403
250mL Storage Flask, Cajon, Airfree, Schlenk, 0-4mm Valve, 160mm OAH. Storage flask sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.