Learn More
Chemglass Life Sciences 25mL Storage Vessel, #15 O-Ring jt, Airfree, Schlenk, 0-4mm Valve, 175mm OAH
Supplier: Chemglass Life Sciences AF009201
25mL Storage Vessel, #15 O-Ring jt, Airfree, Schlenk, 0-4mm Valve, 175mm OAH. Storage vessel with a #15 o-ring joint on the sidearm and a Chem-Vac™ Chem-Cap valve to control gas or liquid flow into the vessel. Vessels are constructed of medium wall tubing and are used with AF-0060 and AF-0061 manifolds or any other Airfree component with a #15 o-ring joint. Supplied complete with one #116 Viton o-ring.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.