Learn More
Essendant AquaDozer Heavy-Duty Squeegee, Black Rubber, Straight, 24" Wide Blade
Supplier: Essendant UNGFP600
Sturdy construction with 14-gauge powder-coated steel frame and reinforced zinc alloy handle socket. Use for moving large volumes of water, mud, debris, waste, scraps, slush, snow and flood clean-up on coated and uncoated smooth concrete floors, asphalt and other smooth surfaces. Great for construction sites, plants, factories, large commercial areas, loading docks, agricultural, food processing, arenas, outdoor areas and around large drains. Replaceable rubber blade.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.