Learn More
HANNA INSTRUMENTS INC 240 mV @ 25°C ORP Test Solution (500 mL)
Supplier: HANNA INSTRUMENTS INC HI7021L
HI7021L is a quality ORP test solution made from reagent grade chemicals and used to test platinum and gold ORP electrodes. Hanna test solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution.Prepared with reagent grade chemicalsAllows for a quick check of your ORP electrodeAir-tight seal to ensure the quality of the solution
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.