Learn More
Cole-Parmer Fowler 52-265-024 24in/600MM Rigid Pre-Cert. Rule
Supplier: Cole-Parmer 9760459
This 24"/600mm rigid ruler shows graduations in 32nds, 64ths, 1mm, and 0.5mm. Includes NIST-traceable calibration report.