Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 234X Uncoated HPPE Cut-Resistant Gloves
Click to view available options
Size:
2X-Large, 10
Large, 8
Medium, 7
Small, 6
X-Large, 9
Description
- Resilient cut-resistant glove offering comfort and flexibility
- ANSI cut level A4 and EN 388:2016 level D
- Cooling HPPE properties and breathable back of hand reduces perspiration and keeps hands dry
- Designed for use in knife hand applications in food processing and food service industries
- Can be used as a glove liner for added protection and comfort
- Thin and lightweight for enhanced dexterity and longer use
- Launderable for re-use, leads to less waste and cost efficiency
- Seamless knit designed to prevent irritation
Specifications
Certifications/Compliance | FDA Compliant, CE Cat. 2 |
Certifications/Compliance (Europe) | Cat II CE, EN 407 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | Gray |
Cut Resistance Rating | ANSI A4 |
Hand | Ambidextrous |
Latex Free | No |
Material | HPPE, Engineered Yarn |
Product Type | Cut Resistant Gloves |
Show More |