Learn More
Chemglass Life Sciences Glas-col Mantle Tripod Support for Series inMin, Mantle Size 22000mL, 14in Fixed Height, 489mm ID
SDP
Supplier:  Chemglass Life Sciences CG1100604
Encompass_Preferred
Glas-col Mantle Tripod Support for Series inMin, Mantle Size 22000mL, 14in Fixed Height, 489mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled.
Catalog No. CG1100604