Learn More
Chemglass Life Sciences 20mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 25 O.D. x 180mm Height
Supplier: Chemglass Life Sciences AF008403
20mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 25 O.D. x 180mm Height. Round bottom, cylindrical drying ampuls with 10mm OD top stem. Manufactured from Type 1 borosilicate glass. Please note: Ampuls are available with top stems to fit AF-0067 Cajon Unions.