Learn More
Chemglass Life Sciences Reaction Kettle Lid, 6-neck, 200mm
Supplier: Chemglass Life Sciences CG1946C10
Reaction Kettle Lid, 6-neck, 200mm. Six-Neck lid for 200mm flanged vessel. Lid has 45/50 center neck, four 45/50 side necks and a 60mm angled flange for powder addition.PLEASE NOTE: 60MM LID, O-RING AND CLAMP MUST BE ORDERED SEPARATELY.