Learn More
Siglent Technologies Oscilloscope 2CH 200 MHz 7''TF
Supplier: Siglent Technologies SDS1202XE
200MHz Bench, Digital Oscilloscopes Ethernet, USB Interface LCD - Color Display 2 Channel Record, Playback, Save.