Learn More
Chemglass Life Sciences Funnel, 2000mL, Separatory, Blank, Stopper Neck, Size # 38, Glassblowers
Supplier: Chemglass Life Sciences CG62407
Funnel, 2000mL, Separatory, Blank, Stopper Neck, Size # 38, Glassblowers. Squibb shaped separatory blank complete with tooled and finished ground stopper neck top. Lower portion of blank is unfinished and requires a stopcock to be sealed. NOTE: SUPPLIED WITHOUT STOPPER.