Learn More
Chemglass Life Sciences 2000mL Style Freeze Dry Flask, P/N 7543200
Supplier: Chemglass Life Sciences CG161109
2000mL Style Freeze Dry Flask, P/N 7543200. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.