Learn More
Polysciences, Inc. Fluoresbrite YG Microspheres 20.0 um
Supplier: Polysciences, Inc. 190962
Fluoresbrite Yellow Green Microspheres have excitation and emission spectra similar to FITC with excitation maxima of 441nm and emission maxima at 485nm. They are one of the brightest microspheres available at this wavelength and are used extensively in phagocytosis studies, flow cytometry and diagnostic assays.