Learn More
Container & Packaging Supply White PP Plastic 20-410, Sprayer, 1700/cs
Supplier: Container & Packaging Supply S199WH
White PP plastic 20-410 ribbed skirt fine mist fingertip sprayer with clear overcap and 5.3125 inch dip tube (.16 cc output).