Learn More
Chemglass Life Sciences 2in Diameter X 1/16in Thick Quartz Disc
Supplier: Chemglass Life Sciences CGQ060007
2in Diameter X 1/16in Thick Quartz Disc. Commercial quality ground and polished clear fused quartz discs. Edges have a ground finished on all sizes.SIZES OTHER THAN THOSE LISTED ARE AVAILABLE. PLEASE CALL FOR INFORMATION.